Gene Transcripts

Gene Symbol
MEOX2
Entrez Gene ID
4223
Gene Name
mesenchyme homeobox 2
Gene Aliases
GAX, MOX2
Chromosome Location
Chr. 7: 15611212 - 15686683 on Build GRCh38
UniGene ID
Hs.170355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005924.4 NP_005915.2 1, 2 800
GenBank mRNA AK309472.1 - 3, 4 429
AK313386.1 - 1, 2 725
BC017021.1 - 1, 2 697 3917118
CR536553.1 - 1, 2 516
L36328.1 - 1, 2 545
X82629.1 - 1, 2 781

View Full Product Details