Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007792.1 | NP_001007793.1 | 12 | 1448 | |
NM_001012331.1 | NP_001012331.1 | 11 | 1514 | ||
NM_002529.3 | NP_002520.2 | 12 | 1532 | ||
GenBank mRNA | AK126428.1 | - | 11 | 1386 | |
AK290759.1 | - | 11 | 1499 | ||
AK312704.1 | - | 12 | 1517 | ||
BC062580.1 | - | 12 | 1475 | ||
BC136554.1 | - | 11 | 1558 | 9020544 | |
BC144239.1 | - | 11 | 1558 | 9052759 | |
CR936794.1 | - | 13 | 1516 | ||
KJ534902.1 | - | 11 | 1458 | ||
KJ534903.1 | - | 8 | 912 | ||
M23102.1 | - | 11 | 1578 |