Gene Transcripts

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
Chromosome Location
Chr. 11: 66848522 - 66958418 on Build GRCh38
UniGene ID
Hs.89890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000920.3 NP_000911.2 22 3452
NM_001040716.1 NP_001035806.1 23 3640
NM_022172.2 NP_071504.2 21 3407
XM_005274031.4 XP_005274088.1 21 3499
XM_005274032.4 XP_005274089.1 21 3381
XM_006718578.3 XP_006718641.1 21 3452
XM_006718579.3 XP_006718642.1 13 2241
XM_011545086.2 XP_011543388.1 21 3768
XM_011545087.2 XP_011543389.1 13 2366
XM_017017868.1 XP_016873357.1 24 3939
XM_017017869.1 XP_016873358.1 24 3810
XM_017017870.1 XP_016873359.1 23 3751
XM_017017871.1 XP_016873360.1 23 3622
XM_017017872.1 XP_016873361.1 22 4004
GenBank mRNA BC011617.2 - 22 3441
M26122.1 - 2 112
S72370.1 - 21 3440
U04641.1 - 21 3396
U30891.1 - 22 3410

View Full Product Details