Gene Transcripts
- Gene Symbol
- ABCB4
- Entrez Gene ID
- 5244
- Gene Name
- ATP binding cassette subfamily B member 4
- Gene Aliases
- ABC21, GBD1, ICP3, MDR2, MDR2/3, MDR3, PFIC-3, PGY3
- Chromosome Location
- Chr. 7: 87399461 - 87475864 on Build GRCh38
- UniGene ID
- Hs.654403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000443.3 |
NP_000434.1
|
16, 17 |
2135 |
|
NM_018849.2 |
NP_061337.1
|
16, 17 |
2135 |
|
NM_018850.2 |
NP_061338.1
|
16, 17 |
2135 |
|
XM_011516308.2 |
XP_011514610.2
|
16, 17 |
2159 |
|
XM_011516309.2 |
XP_011514611.2
|
16, 17 |
2159 |
|
XM_011516310.2 |
XP_011514612.2
|
16, 17 |
2159 |
|
XM_011516311.2 |
XP_011514613.2
|
16, 17 |
2159 |
|
XM_011516312.2 |
XP_011514614.2
|
16, 17 |
2159 |
|
XM_011516313.2 |
XP_011514615.1
|
16, 17 |
2158 |
|
XM_011516315.2 |
XP_011514617.2
|
11, 12 |
1535 |
|
XM_017012323.1 |
XP_016867812.1
|
16, 17 |
2146 |
|
GenBank mRNA |
BC042531.1 |
- |
16, 17 |
2171 |
|
M23234.1 |
- |
16, 17 |
2091 |
|
View Full Product Details