Gene Transcripts

Gene Symbol
PLRG1
Entrez Gene ID
5356
Gene Name
pleiotropic regulator 1
Gene Aliases
Cwc1, PRL1, PRP46, PRPF46, TANGO4
Chromosome Location
Chr. 4: 154534997 - 154550433 on Build GRCh38
UniGene ID
Hs.249996
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201564.1 NP_001188493.1 7, 8 682
NM_002669.3 NP_002660.1 7, 8 709
GenBank mRNA AF044333.1 - 6, 7 584
AK002108.1 - 7, 8 684
AK304574.1 - 6, 7 577
BC020786.1 - 7, 8 646 4809482
BC106004.1 - 7, 8 647 3889109

View Full Product Details