Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018111.2 | NP_001018121.1 | 2 | 416 | |
NM_005397.3 | NP_005388.2 | 2 | 416 | ||
GenBank mRNA | AK055816.1 | - | 3 | 392 | |
AK223573.1 | - | 2 | 408 | ||
BC042466.1 | - | 2 | 368 | ||
BC093730.1 | - | 2 | 178 | 7939575 | |
BC112035.1 | - | 2 | 190 | 8327503 | |
BC143318.1 | - | 2 | 184 | 9051823 | |
U97519.1 | - | 2 | 414 |