Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000305.2 | NP_000296.2 | 4 | 366 | |
NM_001018161.1 | NP_001018171.1 | 4 | 366 | ||
XM_005250453.1 | XP_005250510.1 | 4 | 296 | ||
XM_017012357.1 | XP_016867846.1 | 3 | 215 | ||
XM_017012358.1 | XP_016867847.1 | 3 | 241 | ||
GenBank mRNA | AB102891.1 | - | 4 | 245 | |
AF001601.1 | - | 4 | 277 | ||
AF001602.1 | - | 4 | 288 | ||
AK291103.1 | - | 4 | 288 | ||
AK296029.1 | - | 4 | 281 | ||
AK296636.1 | - | 4 | 322 | ||
AK301313.1 | - | 4 | 493 | ||
AK308065.1 | - | 4 | 297 | ||
AK315209.1 | - | 4 | 314 | ||
BC009728.2 | - | 4 | 322 | ||
BC040010.1 | - | 4 | 269 | 6056277 | |
BC046160.1 | - | 4 | 285 | 5393700 | |
CB961097.1 | - | 395 | |||
L48513.1 | - | 4 | 262 |