Gene Transcripts
- Gene Symbol
- POU2F1
- Entrez Gene ID
- 5451
- Gene Name
- POU class 2 homeobox 1
- Gene Aliases
- OCT1, OTF1, oct-1B
- Chromosome Location
- Chr. 1: 167220829 - 167427345 on Build GRCh38
- UniGene ID
- Hs.283402
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198783.1 |
NP_001185712.1
|
15, 16 |
2189 |
|
NM_001198786.1 |
NP_001185715.1
|
14, 15 |
1867 |
|
NM_002697.3 |
NP_002688.3
|
15, 16 |
2056 |
|
NR_037163.1 |
|
16, 17 |
2100 |
|
XM_011509653.2 |
XP_011507955.1
|
16, 17 |
2008 |
|
XM_011509654.2 |
XP_011507956.1
|
15, 16 |
1969 |
|
GenBank mRNA |
AK091438.1 |
- |
17, 18 |
2484 |
|
AK303201.1 |
- |
14, 15 |
1805 |
|
AY113189.1 |
- |
15, 16 |
2008 |
|
BC001664.2 |
- |
15, 16 |
1972 |
2966289 |
BC003571.1 |
- |
15, 16 |
1972 |
3544378 |
BC052274.1 |
- |
16, 17 |
2080 |
4622256 |
KT438684.1 |
- |
17, 18 |
2167 |
|
X13403.1 |
- |
15, 16 |
1977 |
|
View Full Product Details