Gene Transcripts

Gene Symbol
PPID
Entrez Gene ID
5481
Gene Name
peptidylprolyl isomerase D
Gene Aliases
CYP-40, CYPD
Chromosome Location
Chr. 4: 158709127 - 158723400 on Build GRCh38
UniGene ID
Hs.183958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005038.2 NP_005029.1 5 738
GenBank mRNA AK313929.1 - 5 739
BC030707.2 - 5 734 4819748
CR542081.1 - 5 626
L11667.1 - 5 725

View Full Product Details