Gene Transcripts

Gene Symbol
PPOX
Entrez Gene ID
5498
Gene Name
protoporphyrinogen oxidase
Gene Aliases
PPO, V290M, VP
Chromosome Location
Chr. 1: 161165728 - 161178277 on Build GRCh38
UniGene ID
Hs.517373
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000309.3 NP_000300.1 5 655
NM_001122764.1 NP_001116236.1 5 716
XM_005245291.3 XP_005245348.2 5 694
XM_005245295.3 XP_005245352.2 4 523
XM_006711402.2 XP_006711465.2 5 661
XM_006711403.2 XP_006711466.2 5 650
XM_006711404.3 XP_006711467.2 5 598
XM_006711406.2 XP_006711469.2 5 552
XM_011509663.2 XP_011507965.1 5 781
XM_011509664.1 XP_011507966.1 5 552
XM_011509665.2 XP_011507967.1 5 779
XM_011509666.2 XP_011507968.1 5 786
XM_011509667.2 XP_011507969.1 5 722
XM_011509668.2 XP_011507970.1 5 522
XM_011509669.1 XP_011507971.1 5 535
XM_011509671.1 XP_011507973.1 5 552
XM_011509674.2 XP_011507976.1 5 787
XM_011509676.1 XP_011507978.1 4 847
XM_011509677.1 XP_011507979.1 4 780
XM_011509678.1 XP_011507980.1 5 740
XM_011509679.1 XP_011507981.1 5 548
XM_011509682.1 XP_011507984.1 5 639
XM_017001559.1 XP_016857048.1 5 778
XM_017001560.1 XP_016857049.1 5 694
XM_017001561.1 XP_016857050.1 4 1187
XM_017001562.1 XP_016857051.1 5 621
XM_017001563.1 XP_016857052.1 5 738
XM_017001565.1 XP_016857054.1 5 621
XM_017001566.1 XP_016857055.1 5 730
XM_017001567.1 XP_016857056.1 4 523
XM_017001568.1 XP_016857057.1 5 730
GenBank mRNA BC002357.2 - 5 636
BX384814.2 - 5 729
D38537.1 - 5 703
U26446.1 - 5 617

View Full Product Details