Gene Transcripts

Gene Symbol
PYGB
Entrez Gene ID
5834
Gene Name
phosphorylase, glycogen; brain
Gene Aliases
GPBB
Chromosome Location
Chr. 20: 25248070 - 25298012 on Build GRCh38
UniGene ID
Hs.368157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002862.3 NP_002853.2 3, 4 543
GenBank mRNA AB209080.1 - 3, 4 501
AK299697.1 - 3, 4 467
AK309036.1 - 3, 4 541
BC017045.2 - 3, 4 518 3877951
BC030795.2 - 3, 4 519 3916938
BQ433140.1 - 3, 4 543
J03544.1 - 3, 4 513
U47025.1 - 3, 4 468

View Full Product Details