Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002888.3 | NP_002879.2 | 4 | 637 | |
NM_206963.1 | NP_996846.1 | 4 | 637 | ||
XM_005247686.4 | XP_005247743.3 | 4 | 1053 | ||
GenBank mRNA | AK130079.1 | - | 4 | 608 | |
AW514087.1 | - | 1 | 246 | ||
BC029640.1 | - | 4 | 573 | 4776506 | |
BM919188.1 | - | 4 | 764 | ||
U27185.1 | - | 4 | 602 |