Gene Transcripts
- Gene Symbol
- SHBG
- Entrez Gene ID
- 6462
- Gene Name
- sex hormone binding globulin
- Gene Aliases
- ABP, SBP, TEBG
- Chromosome Location
- Chr. 17: 7614064 - 7633383 on Build GRCh38
- UniGene ID
- Hs.632235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040.4 |
NP_001031.2
|
7, 8 |
1146 |
|
NM_001146279.2 |
NP_001139751.1
|
7, 8 |
1092 |
|
NM_001289113.1 |
NP_001276042.1
|
7, 8 |
1139 |
|
NM_001289114.1 |
NP_001276043.1
|
7, 8 |
1002 |
|
NM_001289116.1 |
NP_001276045.1
|
7, 8 |
1054 |
|
GenBank mRNA |
BC069597.1 |
- |
7, 8 |
1064 |
|
BC101785.1 |
- |
7, 8 |
1098 |
8069291 |
BC112186.1 |
- |
7, 8 |
1098 |
8327654 |
CD013957.1 |
- |
7, 8 |
1034 |
|
CD013958.1 |
- |
7, 8 |
996 |
|
EU352664.1 |
- |
3, 4 |
319 |
|
EU352665.1 |
- |
7, 8 |
1007 |
|
EU352669.1 |
- |
3, 4 |
340 |
|
EU352670.1 |
- |
7, 8 |
1002 |
|
EU352674.1 |
- |
3, 4 |
335 |
|
EU352675.1 |
- |
6, 7 |
840 |
|
X05403.1 |
- |
7, 8 |
1004 |
|
X05792.1 |
- |
5, 6 |
709 |
|
X05885.1 |
- |
6, 7 |
931 |
|
View Full Product Details