Gene Transcripts

Gene Symbol
SHOX
Entrez Gene ID
6473
Gene Name
short stature homeobox
Gene Aliases
GCFX, PHOG, SHOXY, SS
Chromosome Location
Chr. Y: 624344 - 659411 on Build GRCh38
UniGene ID
Hs.105932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000451.3 NP_000442.1 4 1219
NM_006883.2 NP_006874.1 4 1219
GenBank mRNA U89331.1 - 4 1221
Y11535.1 - 3 619
Y11536.1 - 3 619

View Full Product Details