Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297774.1 | NP_001284703.1 | 2 | 568 | |
NM_003180.2 | NP_003171.2 | 4 | 620 | ||
XM_006723339.3 | XP_006723402.1 | 4 | 455 | ||
XM_006723340.3 | XP_006723403.1 | 4 | 709 | ||
XM_006723341.3 | XP_006723404.1 | 2 | 1039 | ||
XM_017027175.1 | XP_016882664.1 | 4 | 517 | ||
XM_017027176.1 | XP_016882665.1 | 4 | 508 | ||
GenBank mRNA | AK125182.1 | - | 3 | 1408 | |
AK295337.1 | - | 2 | 515 | ||
AK313067.1 | - | 4 | 409 | ||
BC029962.1 | - | 4 | 548 | ||
BC033683.1 | - | 4 | 427 | ||
BQ417474.1 | - | 4 | 533 | ||
DQ090951.1 | - | 3 | 339 | ||
Y19236.1 | - | 2 | 138 |