Gene Transcripts

Gene Symbol
TBX2
Entrez Gene ID
6909
Gene Name
T-box 2
Gene Aliases
Chromosome Location
Chr. 17: 61399896 - 61409466 on Build GRCh38
UniGene ID
Hs.531085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005994.3 NP_005985.3 2 829
GenBank mRNA AB209378.1 - 2 360
AL832900.1 - 2 369
BC052566.1 - 2 829 6339405
BC070054.1 - 2 794
S81264.1 - 1 65
U28049.1 - 2 565

View Full Product Details