Gene Transcripts

Gene Symbol
TTC1
Entrez Gene ID
7265
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
TPR1
Chromosome Location
Chr. 5: 160009100 - 160065545 on Build GRCh38
UniGene ID
Hs.519718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282500.1 NP_001269429.1 8 956
NM_003314.2 NP_003305.1 8 959
GenBank mRNA AK128840.1 - 1 3610
AK222641.1 - 8 910
AK222984.1 - 8 904
AK315262.1 - 8 914
BC000942.3 - 7 864 3446109
BQ017702.1 - 1 537
U46570.1 - 8 886

View Full Product Details