Gene Transcripts

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
Chromosome Location
Chr. 2: 71453155 - 71686763 on Build GRCh38
UniGene ID
Hs.252180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130455.1 NP_001123927.1 33 3832
NM_001130976.1 NP_001124448.1 32 4022
NM_001130977.1 NP_001124449.1 32 4022
NM_001130978.1 NP_001124450.1 33 4064
NM_001130979.1 NP_001124451.1 34 4157
NM_001130980.1 NP_001124452.1 33 4115
NM_001130981.1 NP_001124453.1 33 4115
NM_001130982.1 NP_001124454.1 34 3925
NM_001130983.1 NP_001124455.1 33 3832
NM_001130984.1 NP_001124456.1 32 3790
NM_001130985.1 NP_001124457.1 33 3883
NM_001130986.1 NP_001124458.1 32 3790
NM_001130987.1 NP_001124459.1 33 3883
NM_003494.3 NP_003485.1 33 4064
XM_005264584.4 XP_005264641.1 34 3940
XM_005264585.4 XP_005264642.1 34 4625
GenBank mRNA AF075575.1 - 33 4061
AK074104.1 - 19 2334
AK295004.1 - 4 521
BC171803.1 - 33 4037
DQ267935.1 - 33 3832
EU515155.1 - 34 4058
EU515156.1 - 32 3923
EU515157.1 - 33 3965
EU515158.1 - 33 4016
EU515159.1 - 32 3923
EU515160.1 - 33 4016
EU515161.1 - 34 3925
EU515162.1 - 32 3790
EU515163.1 - 33 3832
EU515164.1 - 33 3883
EU515165.1 - 32 3790
EU515166.1 - 33 3883

View Full Product Details