Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007226.1 | NP_001007227.1 | 6 | 806 | |
NM_001007227.1 | NP_001007228.1 | 5 | 684 | ||
NM_001007228.1 | NP_001007229.1 | 4 | 632 | ||
NM_001007229.1 | NP_001007230.1 | 5 | 683 | ||
NM_001007230.1 | NP_001007231.1 | 6 | 735 | ||
NM_003563.3 | NP_003554.1 | 5 | 754 | ||
XM_005257723.4 | XP_005257780.1 | 5 | 978 | ||
XM_005257724.4 | XP_005257781.1 | 4 | 908 | ||
XM_017025204.1 | XP_016880693.1 | 3 | 421 | ||
GenBank mRNA | AJ000644.1 | - | 4 | 492 | |
AK222589.1 | - | 4 | 457 | ||
AK293045.1 | - | 5 | 965 | ||
AK300616.1 | - | 4 | 456 | ||
AK303268.1 | - | 3 | 378 | ||
AK312691.1 | - | 4 | 468 | ||
AK316366.1 | - | 6 | 533 | ||
BC001269.1 | - | 5 | 638 | 3461565 | |
BC003385.1 | - | 4 | 449 | 3445878 |