Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142930.1 | NP_001136402.1 | 1 | 189 | |
NM_001142931.1 | NP_001136403.1 | 1 | 189 | ||
NM_001243747.1 | NP_001230676.1 | 1 | 189 | ||
NM_006595.3 | NP_006586.1 | 1 | 189 | ||
NR_024625.1 | 1 | 189 | |||
XM_006718359.3 | XP_006718422.1 | 1 | 179 | ||
XM_017018464.1 | XP_016873953.1 | 1 | 174 | ||
XM_017018465.1 | XP_016873954.1 | 1 | 174 | ||
GenBank mRNA | AF229254.1 | - | 1 | 129 | |
AK293303.1 | - | 1 | 90 | ||
AK294724.1 | - | 1 | 146 | ||
AK299288.1 | - | 1 | 155 | ||
AK304157.1 | - | 1 | 155 | ||
AY311389.1 | - | 1 | 128 | ||
BC017709.1 | - | 1 | 109 | 4419401 | |
BM193468.1 | - | 1 | 171 | ||
BT007093.1 | - | 1 | 16 | ||
DA104687.1 | - | 1 | 190 | ||
DC334531.1 | - | 1 | 189 | ||
U83857.1 | - | 1 | 111 | ||
Y15906.1 | - | 1 | 148 |