Gene Transcripts

Gene Symbol
MTMR1
Entrez Gene ID
8776
Gene Name
myotubularin related protein 1
Gene Aliases
Chromosome Location
Chr. X: 150692997 - 150765103 on Build GRCh38
UniGene ID
Hs.347187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306144.1 NP_001293073.1 12 1569
NM_003828.3 NP_003819.1 11 1545
XM_005274765.3 XP_005274822.1 14 1609
XM_005274766.3 XP_005274823.1 13 1557
XM_005274767.3 XP_005274824.1 12 1576
XM_006724855.3 XP_006724918.1 11 1545
XM_011531209.2 XP_011529511.1 11 1305
XM_017029920.1 XP_016885409.1 14 1609
XM_017029921.1 XP_016885410.1 12 1340
XM_017029922.1 XP_016885411.1 12 1352
XM_017029923.1 XP_016885412.1 9 1132
GenBank mRNA AF057354.1 - 11 1369
AJ224979.1 - 11 1371
AK001816.1 - 8 888
AK023451.1 - 2 1518
AK226109.1 - 12 1523
AK295738.1 - 12 1364
U58032.1 - 8 1150

View Full Product Details