Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003845.2 | NP_003836.1 | 7 | 821 | |
GenBank mRNA | AF263541.1 | - | 6 | 721 | |
AK292322.1 | - | 7 | 815 | ||
AK308260.1 | - | 9 | 1042 | ||
BC031244.1 | - | 7 | 815 | 5266287 | |
GU139743.1 | - | 6 | 805 |