Gene Transcripts

Gene Symbol
TAX1BP1
Entrez Gene ID
8887
Gene Name
Tax1 binding protein 1
Gene Aliases
CALCOCO3, T6BP, TXBP151
Chromosome Location
Chr. 7: 27739373 - 27829767 on Build GRCh38
UniGene ID
Hs.34576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079864.2 NP_001073333.1 9 1360
NM_001206901.1 NP_001193830.1 9 1363
NM_001206902.1 NP_001193831.1 8 1172
NM_006024.6 NP_006015.4 9 1360
XM_005249900.2 XP_005249957.1 9 1360
XM_017012754.1 XP_016868243.1 9 1360
GenBank mRNA AF090891.1 - 9 1278
AF268075.1 - 8 1178
AK000098.1 - 9 1273
AK296720.1 - 8 1121
AK299729.1 - 9 1363
AK301064.1 - 9 1149
AK314292.1 - 9 1330
AL136586.1 - 9 1266
BC050358.2 - 9 1245 6055189
CR457056.1 - 8 1178
CR533556.1 - 8 1178
U33821.2 - 9 1261

View Full Product Details