Gene Transcripts

Gene Symbol
BSN
Entrez Gene ID
8927
Gene Name
bassoon presynaptic cytomatrix protein
Gene Aliases
ZNF231
Chromosome Location
Chr. 3: 49554489 - 49671549 on Build GRCh38
UniGene ID
Hs.194684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003458.3 NP_003449.2 5 2937
GenBank mRNA AF052224.1 - 5 2938

View Full Product Details