Gene Transcripts
- Gene Symbol
- DNAJA3
- Entrez Gene ID
- 9093
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A3
- Gene Aliases
- HCA57, TID1, hTID-1
- Chromosome Location
- Chr. 16: 4425805 - 4456775 on Build GRCh38
- UniGene ID
- Hs.459779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135110.2 |
NP_001128582.1
|
4, 5 |
695 |
|
NM_001286516.1 |
NP_001273445.1
|
2, 3 |
346 |
|
NM_005147.5 |
NP_005138.3
|
4, 5 |
695 |
|
GenBank mRNA |
AB209923.1 |
- |
4, 5 |
614 |
|
AF061749.1 |
- |
4, 5 |
649 |
|
AF244136.1 |
- |
4, 5 |
620 |
|
AF411044.1 |
- |
4, 5 |
662 |
|
AK001333.1 |
- |
4, 5 |
635 |
|
AK223105.1 |
- |
4, 5 |
635 |
|
AK295391.1 |
- |
2, 3 |
286 |
|
AK314218.1 |
- |
4, 5 |
632 |
|
BC007225.1 |
- |
4, 5 |
615 |
|
BC011855.2 |
- |
4, 5 |
619 |
4139881 |
BC012343.1 |
- |
4, 5 |
611 |
4653507 |
BC014062.2 |
- |
4, 5 |
611 |
|
BC030145.2 |
- |
4, 5 |
611 |
|
BC032100.2 |
- |
4, 5 |
633 |
4660602 |
BX445320.2 |
- |
4, 5 |
695 |
|
View Full Product Details