Gene Transcripts

Gene Symbol
WDR46
Entrez Gene ID
9277
Gene Name
WD repeat domain 46
Gene Aliases
BING4, C6orf11, FP221, UTP7
Chromosome Location
Chr. 6: 33278850 - 33289527 on Build GRCh38
UniGene ID
Hs.520063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164267.1 NP_001157739.1 6 797
NM_005452.5 NP_005443.3 6 959
XM_017011484.1 XP_016866973.1 6 673
XM_017011485.1 XP_016866974.1 4 321
XM_017011486.1 XP_016866975.1 3 176
XM_017011487.1 XP_016866976.1 4 321
GenBank mRNA AF447870.1 - 6 651
AK292171.1 - 6 942
AK292616.1 - 6 742
AK297925.1 - 6 554
AK298151.1 - 6 508
AK308534.1 - 5 435
AK311388.1 - 5 656
AK315616.1 - 6 671
BC000388.2 - 6 646 2821198
BX439421.2 - 6 646

View Full Product Details