Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004798.3 | NP_004789.1 | 5 | 1914 | |
GenBank mRNA | AB002357.1 | - | 5 | 1901 | |
AK098431.1 | - | 4 | 459 | ||
AK299877.1 | - | 5 | 837 | ||
BC136310.1 | - | 4 | 1807 | 9020297 | |
BC136311.1 | - | 4 | 1807 | 9020298 |