Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004869.3 | NP_004860.2 | 5 | 714 | |
XM_006722582.2 | XP_006722645.1 | 4 | 590 | ||
XM_011526268.2 | XP_011524570.1 | 5 | 714 | ||
GenBank mRNA | AF038960.1 | - | 5 | 643 | |
AF195514.1 | - | 5 | 655 | ||
AK290932.1 | - | 5 | 658 | ||
AK291253.1 | - | 5 | 612 | ||
AK308892.1 | - | 5 | 678 | ||
AK309322.1 | - | 4 | 590 | ||
AK311041.1 | - | 4 | 590 | ||
AY232629.1 | - | 5 | 464 | ||
BC039574.1 | - | 5 | 621 | 6042862 |