Gene Transcripts

Gene Symbol
SNCAIP
Entrez Gene ID
9627
Gene Name
synuclein alpha interacting protein
Gene Aliases
SYPH1, Sph1
Chromosome Location
Chr. 5: 122311354 - 122464219 on Build GRCh38
UniGene ID
Hs.426463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308100.1 NP_001295029.1 6 2167
NM_001308105.1 NP_001295034.1 3 506
NM_005460.3 NP_005451.2 4 722
NR_131761.1 4 647
XM_005272138.3 XP_005272195.1 4 645
XM_005272139.1 XP_005272196.1 4 585
XM_006714734.2 XP_006714797.1 4 778
XM_011543737.2 XP_011542039.1 5 780
XM_011543738.2 XP_011542040.1 5 779
XM_011543739.1 XP_011542041.1 5 705
XM_011543741.2 XP_011542043.1 5 729
XM_011543743.2 XP_011542045.1 5 1154
XM_011543745.1 XP_011542047.1 4 609
XM_017010078.1 XP_016865567.1 5 3107
XM_017010079.1 XP_016865568.1 5 780
XM_017010080.1 XP_016865569.1 5 780
XM_017010081.1 XP_016865570.1 5 778
XM_017010082.1 XP_016865571.1 4 609
XM_017010084.1 XP_016865573.1 5 786
XM_017010086.1 XP_016865575.1 5 784
GenBank mRNA AB110788.1 - 3 460
AB110789.1 - 3 460
AB110790.1 - 4 601
AF076929.1 - 4 553
AK310835.1 - 4 554
BC040552.1 - 6 2029 5298588

View Full Product Details