Gene Transcripts
- Gene Symbol
- TOM1
- Entrez Gene ID
- 10043
- Gene Name
- target of myb1 membrane trafficking protein
- Gene Aliases
- Chromosome Location
- Chr. 22: 35299275 - 35347994 on Build GRCh38
- UniGene ID
- Hs.474705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135730.1 |
NP_001129202.1
|
13, 14 |
1311 |
|
NM_005488.2 |
NP_005479.1
|
14, 15 |
1446 |
|
NR_024194.1 |
|
14, 15 |
1475 |
|
NR_024195.1 |
|
13, 14 |
1396 |
|
XM_017028529.1 |
XP_016884018.1
|
15, 16 |
1460 |
|
XM_017028530.1 |
XP_016884019.1
|
13, 14 |
1311 |
|
GenBank mRNA |
AJ006973.1 |
- |
14, 15 |
1382 |
|
AK026576.1 |
- |
14, 15 |
1399 |
|
AK090723.1 |
- |
2, 3 |
955 |
|
AK091958.1 |
- |
2, 3 |
1637 |
|
AK097066.1 |
- |
12, 13 |
957 |
|
AK097926.1 |
- |
16, 17 |
1662 |
|
AK295918.1 |
- |
13, 14 |
1306 |
|
AK296670.1 |
- |
14, 15 |
1383 |
|
AK297827.1 |
- |
13, 14 |
1225 |
|
AK314522.1 |
- |
14, 15 |
1355 |
|
BC040006.1 |
- |
14, 15 |
1339 |
|
CR456595.1 |
- |
14, 15 |
1342 |
|
View Full Product Details