Gene Transcripts
- Gene Symbol
- TOM1
- Entrez Gene ID
- 10043
- Gene Name
- target of myb1 membrane trafficking protein
- Gene Aliases
- Chromosome Location
- Chr. 22: 35299275 - 35347994 on Build GRCh38
- UniGene ID
- Hs.474705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135729.1 |
NP_001129201.1
|
8, 9 |
954 |
|
NM_001135730.1 |
NP_001129202.1
|
7, 8 |
892 |
|
NM_001135732.1 |
NP_001129204.1
|
8, 9 |
1027 |
|
NM_005488.2 |
NP_005479.1
|
8, 9 |
1027 |
|
NR_024194.1 |
|
8, 9 |
1056 |
|
NR_024195.1 |
|
7, 8 |
977 |
|
XM_011529818.2 |
XP_011528120.1
|
9, 10 |
1041 |
|
XM_011529820.2 |
XP_011528122.1
|
7, 8 |
892 |
|
XM_017028529.1 |
XP_016884018.1
|
9, 10 |
1041 |
|
XM_017028530.1 |
XP_016884019.1
|
7, 8 |
892 |
|
GenBank mRNA |
AJ006973.1 |
- |
8, 9 |
963 |
|
AK026576.1 |
- |
8, 9 |
980 |
|
AK097066.1 |
- |
6, 7 |
538 |
|
AK293694.1 |
- |
8, 9 |
954 |
|
AK295918.1 |
- |
7, 8 |
887 |
|
AK296670.1 |
- |
8, 9 |
964 |
|
AK297827.1 |
- |
7, 8 |
806 |
|
AK314522.1 |
- |
8, 9 |
936 |
|
BC040006.1 |
- |
8, 9 |
920 |
|
BC046151.1 |
- |
8, 9 |
905 |
6470390 |
CR456595.1 |
- |
8, 9 |
923 |
|
View Full Product Details