Gene Transcripts

Gene Symbol
MAEA
Entrez Gene ID
10296
Gene Name
macrophage erythroblast attacher
Gene Aliases
EMLP, EMP, GID9, HLC-10, PIG5
Chromosome Location
Chr. 4: 1289851 - 1340148 on Build GRCh38
UniGene ID
Hs.139896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017405.2 NP_001017405.1 5 687
NM_001297430.1 NP_001284359.1 4 483
NM_001297432.1 NP_001284361.1 5 825
NM_001297433.1 NP_001284362.1 5 761
NM_005882.4 NP_005873.2 4 564
XM_006713849.3 XP_006713912.1 5 647
XM_006713850.2 XP_006713913.1 5 820
XM_011513376.1 XP_011511678.1 6 807
XM_011513377.1 XP_011511679.1 5 725
XM_017007636.1 XP_016863125.1 6 807
XM_017007637.1 XP_016863126.1 5 724
GenBank mRNA AF084928.1 - 5 512
AK001088.1 - 5 647
AK022586.1 - 4 524
AK296174.1 - 4 440
AK298944.1 - 5 741
AK300572.1 - 6 584
AK301155.1 - 5 655
AK308901.1 - 4 508
AY189687.1 - 5 600
BC001225.1 - 5 654 3533990
BC006470.1 - 5 610
BT006957.1 - 5 591

View Full Product Details