Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019112.3 | NP_061985.2 | 30 | 4395 | |
XM_006722616.1 | XP_006722679.1 | 30 | 4382 | ||
XM_006722617.2 | XP_006722680.1 | 30 | 4382 | ||
XM_006722618.2 | XP_006722681.1 | 15 | 1932 | ||
XM_011527628.2 | XP_011525930.1 | 30 | 4256 | ||
XM_011527629.1 | XP_011525931.1 | 30 | 4355 | ||
XM_011527630.1 | XP_011525932.1 | 30 | 4382 | ||
XM_011527631.1 | XP_011525933.1 | 30 | 4382 | ||
XM_011527632.1 | XP_011525934.1 | 26 | 3834 | ||
XM_011527633.2 | XP_011525935.1 | 30 | 4382 | ||
XM_011527634.1 | XP_011525936.1 | 30 | 4382 | ||
XM_011527635.1 | XP_011525937.1 | 30 | 4382 | ||
XM_011527636.2 | XP_011525938.1 | 15 | 1930 | ||
XM_017026143.1 | XP_016881632.1 | 30 | 4382 | ||
GenBank mRNA | AB055390.1 | - | 24 | 3750 | |
AF250238.1 | - | 29 | 4164 | ||
AF328787.1 | - | 29 | 4280 |