Gene Transcripts

Gene Symbol
CXCL13
Entrez Gene ID
10563
Gene Name
C-X-C motif chemokine ligand 13
Gene Aliases
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13
Chromosome Location
Chr. 4: 77511753 - 77611834 on Build GRCh38
UniGene ID
Hs.100431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006419.2 NP_006410.1 2, 3 152
XM_006714063.3 XP_006714126.1 1, 2 145
GenBank mRNA AF029894.1 - 1, 2 108
AF044197.1 - 2, 3 164
AJ002211.1 - 1, 2 108
BC012589.1 - 1, 2 124 4246053
BI836728.1 - 1, 2 104
CR407624.1 - 1, 2 74

View Full Product Details