Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006453.2 | NP_006444.2 | 15, 16 | 1765 | |
GenBank mRNA | AB209173.1 | - | 15, 16 | 1737 | |
AK095076.1 | - | 7, 8 | 1941 | ||
BC007620.1 | - | 7, 8 | 949 | ||
BC010231.1 | - | 15, 16 | 1765 | 3455740 | |
BC014007.2 | - | 15, 16 | 1737 | 4040312 | |
BC035409.1 | - | 15, 16 | 1721 | 5165833 | |
U02609.1 | - | 15, 16 | 1712 |