Gene Transcripts
- Gene Symbol
- SDCCAG3
- Entrez Gene ID
- 10807
- Gene Name
- serologically defined colon cancer antigen 3
- Gene Aliases
- NY-CO-3
- Chromosome Location
- Chr. 9: 136401922 - 136410602 on Build GRCh38
- UniGene ID
- Hs.94300
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039707.1 |
NP_001034796.1
|
5, 6 |
1016 |
|
NM_001039708.1 |
NP_001034797.1
|
3, 4 |
797 |
|
NM_006643.3 |
NP_006634.3
|
4, 5 |
947 |
|
XM_005266050.3 |
XP_005266107.1
|
5, 6 |
970 |
|
XM_005266051.3 |
XP_005266108.1
|
4, 5 |
861 |
|
XM_011518157.2 |
XP_011516459.1
|
4, 5 |
800 |
|
XM_017014218.1 |
XP_016869707.1
|
4, 5 |
1117 |
|
XM_017014219.1 |
XP_016869708.1
|
4, 5 |
1117 |
|
GenBank mRNA |
AF039688.1 |
- |
1, 2 |
84 |
|
AK001296.1 |
- |
3, 4 |
576 |
|
AY349357.1 |
- |
4, 5 |
637 |
|
BC014515.1 |
- |
4, 5 |
814 |
3547130 |
View Full Product Details