Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138964.2 | NP_620414.1 | 2 | 520 | |
GenBank mRNA | AB084080.1 | - | 2 | 520 | |
AF506287.1 | - | 2 | 520 | ||
AY089976.1 | - | 2 | 520 | ||
BC100960.2 | - | 2 | 520 | 40004720 | |
EF577399.1 | - | 2 | 520 |