Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159629.1 | NP_001153101.1 | 1, 2 | 716 | |
NM_003645.3 | NP_003636.2 | 1, 2 | 716 | ||
GenBank mRNA | AF096290.1 | - | 1, 2 | 688 | |
AK222859.1 | - | 1, 2 | 687 | ||
AK223145.1 | - | 1, 2 | 685 | ||
AK290262.1 | - | 1, 2 | 716 | ||
BC057770.1 | - | 1, 2 | 679 | 30348317 | |
D88308.1 | - | 1, 2 | 706 |