Gene Transcripts

Gene Symbol
RAB35
Entrez Gene ID
11021
Gene Name
RAB35, member RAS oncogene family
Gene Aliases
H-ray, RAB1C, RAY
Chromosome Location
Chr. 12: 120095094 - 120116839 on Build GRCh38
UniGene ID
Hs.524788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167606.1 NP_001161078.1 2 258
NM_006861.6 NP_006852.1 2 258
XM_011537779.1 XP_011536081.1 2 195
XM_017018723.1 XP_016874212.1 2 160
GenBank mRNA AF498960.1 - 2 69
AK125543.1 - 3 1033
AK300127.1 - 2 176
AK304620.1 - 2 165
AK312538.1 - 2 168
BC015931.2 - 2 173 3907209
BM467132.1 - 2 239
BQ233709.1 - 2 214
BT020024.1 - 2 69
CR536486.1 - 2 69
CR541683.1 - 2 69
DC349830.1 - 2 255
DC370610.1 - 2 258
DC408044.1 - 2 258
X79781.1 - 2 150

View Full Product Details