Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278439.1 | NP_001265368.1 | 7 | 582 | |
NM_007056.2 | NP_008987.2 | 8 | 768 | ||
NR_103529.1 | 8 | 768 | |||
XM_011526396.2 | XP_011524698.1 | 8 | 763 | ||
XM_011526397.2 | XP_011524699.1 | 7 | 597 | ||
GenBank mRNA | AF042800.1 | - | 8 | 768 | |
AK094681.1 | - | 7 | 1362 | ||
AK293333.1 | - | 7 | 578 | ||
AY358944.1 | - | 8 | 749 | ||
BC080554.1 | - | 7 | 708 | 6258057 | |
BC131496.1 | - | 7 | 610 |