Gene Transcripts

Gene Symbol
AKAP11
Entrez Gene ID
11215
Gene Name
A-kinase anchoring protein 11
Gene Aliases
AKAP-11, AKAP220, PPP1R44, PRKA11
Chromosome Location
Chr. 13: 42271470 - 42323267 on Build GRCh38
UniGene ID
Hs.105105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016248.3 NP_057332.1 8 3514
XM_005266247.1 XP_005266304.1 8 3515
XM_005266248.3 XP_005266305.1 7 3408
XM_005266249.3 XP_005266306.1 8 3515
XM_005266250.3 XP_005266307.1 8 3515
XM_011534903.2 XP_011533205.1 8 3511
XM_011534904.1 XP_011533206.1 8 3707
XM_011534905.1 XP_011533207.1 7 3465
XM_011534906.2 XP_011533208.1 7 3683
XM_017020381.1 XP_016875870.1 8 3565
XM_017020382.1 XP_016875871.1 6 3355
XM_017020383.1 XP_016875872.1 6 3543
GenBank mRNA AB014529.2 - 1 1454
AF176555.1 - 8 3514

View Full Product Details