Gene Transcripts

Gene Symbol
EXOC3
Entrez Gene ID
11336
Gene Name
exocyst complex component 3
Gene Aliases
SEC6, SEC6L1, Sec6p
Chromosome Location
Chr. 5: 443184 - 471938 on Build GRCh38
UniGene ID
Hs.481464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007277.4 NP_009208.2 5, 6 1294
XM_011513948.2 XP_011512250.1 5, 6 1329
XM_011513949.2 XP_011512251.1 5, 6 1346
XM_011513950.2 XP_011512252.1 4, 5 1224
XM_017009000.1 XP_016864489.1 5, 6 1329
XM_017009001.1 XP_016864490.1 5, 6 1329
XM_017009002.1 XP_016864491.1 5, 6 1329
XM_017009003.1 XP_016864492.1 5, 6 1329
XM_017009004.1 XP_016864493.1 6, 7 1407
XM_017009006.1 XP_016864495.1 5, 6 1323
GenBank mRNA AF055006.1 - 2, 3 346
AK131103.1 - 2, 3 775
AK315749.1 - 2, 3 802
AL832434.1 - 3, 4 950
BC001511.2 - 2, 3 706
BC019304.1 - 3, 4 876
BC064569.1 - 4, 5 1236 5590332
GU727644.1 - 5, 6 1294

View Full Product Details