Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014939.3 | NP_055754.2 | 5 | 1030 | |
XM_005258233.2 | XP_005258290.1 | 5 | 1120 | ||
XM_005258234.2 | XP_005258291.1 | 5 | 1120 | ||
XM_006722420.3 | XP_006722483.1 | 5 | 1120 | ||
XM_011525884.2 | XP_011524186.1 | 5 | 1120 | ||
XM_017025615.1 | XP_016881104.1 | 5 | 622 | ||
XM_017025616.1 | XP_016881105.1 | 5 | 1120 | ||
XM_017025617.1 | XP_016881106.1 | 5 | 1120 | ||
GenBank mRNA | AB023229.1 | - | 5 | 723 | |
AK292537.1 | - | 5 | 1009 | ||
AL136749.1 | - | 6 | 817 | ||
BC059380.1 | - | 5 | 1009 | ||
BC127109.1 | - | 5 | 584 | 40124744 | |
BC127110.1 | - | 5 | 584 | 40124745 | |
BX642999.1 | - | 5 | 576 |