Gene Transcripts
- Gene Symbol
- SIRT2
- Entrez Gene ID
- 22933
- Gene Name
- sirtuin 2
- Gene Aliases
- SIR2, SIR2L, SIR2L2
- Chromosome Location
- Chr. 19: 38878555 - 38899862 on Build GRCh38
- UniGene ID
- Hs.466693
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193286.1 |
NP_001180215.1
|
2, 3 |
414 |
|
NM_012237.3 |
NP_036369.2
|
3, 4 |
422 |
|
NM_030593.2 |
NP_085096.1
|
2, 3 |
414 |
|
NR_034146.1 |
|
1, 2 |
345 |
|
XM_006723111.1 |
XP_006723174.1
|
4, 5 |
370 |
|
XM_011526654.1 |
XP_011524956.1
|
2, 3 |
307 |
|
XM_017026500.1 |
XP_016881989.1
|
2, 3 |
178 |
|
GenBank mRNA |
AB009274.1 |
- |
2, 3 |
138 |
|
AF083107.2 |
- |
3, 4 |
320 |
|
AF095714.1 |
- |
2, 3 |
193 |
|
AF131800.1 |
- |
1, 2 |
69 |
|
AF160214.1 |
- |
2, 3 |
174 |
|
AK025876.1 |
- |
2, 3 |
200 |
|
AK054642.1 |
- |
3, 4 |
259 |
|
AK225824.1 |
- |
2, 3 |
197 |
|
AK290716.1 |
- |
1, 2 |
345 |
|
AK314492.1 |
- |
3, 4 |
176 |
|
AY030277.1 |
- |
2, 3 |
175 |
|
BC003012.1 |
- |
2, 3 |
265 |
2820929 |
BC003547.1 |
- |
2, 3 |
265 |
3536728 |
DA541045.1 |
- |
2, 3 |
415 |
|
DA705079.1 |
- |
3, 4 |
422 |
|
View Full Product Details