Gene Transcripts

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
Chromosome Location
Chr. 7: 51016212 - 51316818 on Build GRCh38
UniGene ID
Hs.99141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287436.1 NP_001274365.1 11 3334
NM_015198.3 NP_056013.2 10 3163
XM_005271750.1 XP_005271807.1 12 3370
XM_005271751.1 XP_005271808.1 11 3325
XM_005271756.1 XP_005271813.1 10 3154
XM_011515234.1 XP_011513536.1 13 3445
XM_011515235.1 XP_011513537.1 12 3400
XM_011515236.1 XP_011513538.1 13 3445
XM_011515237.1 XP_011513539.1 12 3274
XM_011515238.1 XP_011513540.1 13 3445
XM_011515239.1 XP_011513541.1 12 3274
XM_011515240.1 XP_011513542.1 11 3229
XM_011515241.1 XP_011513543.1 7 2449
XM_017011898.1 XP_016867387.1 11 3199
XM_017011899.1 XP_016867388.1 11 3325
XM_017011900.1 XP_016867389.1 10 3154
GenBank mRNA AB014533.1 - 10 3144
AK293883.1 - 4 1543
AK308718.1 - 1 294
AL713786.1 - 2 6618
BC029275.1 - 5 2034
BC045771.1 - 9 2633
BC136441.1 - 11 3249 9020430
BC144099.1 - 10 3078 9052619
BC150263.1 - 10 3144 9007207
CR936689.1 - 9 2654

View Full Product Details