Gene Transcripts

Gene Symbol
ZDHHC17
Entrez Gene ID
23390
Gene Name
zinc finger DHHC-type containing 17
Gene Aliases
HIP14, HIP3, HSPC294, HYPH
Chromosome Location
Chr. 12: 76764074 - 76853701 on Build GRCh38
UniGene ID
Hs.4014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015336.2 NP_056151.2 12, 13 1486
XM_005268750.4 XP_005268807.1 12, 13 1285
XM_011538083.1 XP_011536385.1 12, 13 1350
XM_017019091.1 XP_016874580.1 13, 14 1436
XM_017019092.1 XP_016874581.1 12, 13 1299
XM_017019093.1 XP_016874582.1 12, 13 1248
XM_017019094.1 XP_016874583.1 11, 12 1166
XM_017019095.1 XP_016874584.1 8, 9 878
GenBank mRNA AB023163.1 - 12, 13 1430
AB024494.1 - 12, 13 1976
AB097013.1 - 12, 13 1306
AB097035.1 - 12, 13 1306
AK292866.1 - 12, 13 1302
BC050324.1 - 12, 13 1445 5266097

View Full Product Details