Gene Transcripts

Gene Symbol
ZDHHC17
Entrez Gene ID
23390
Gene Name
zinc finger DHHC-type containing 17
Gene Aliases
HIP14, HIP3, HSPC294, HYPH
Chromosome Location
Chr. 12: 76764074 - 76853701 on Build GRCh38
UniGene ID
Hs.4014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015336.2 NP_056151.2 17 1978
XM_005268750.4 XP_005268807.1 17 1777
XM_011538083.1 XP_011536385.1 17 1842
XM_017019091.1 XP_016874580.1 18 1928
XM_017019092.1 XP_016874581.1 17 1791
XM_017019093.1 XP_016874582.1 17 1740
XM_017019094.1 XP_016874583.1 16 1658
XM_017019095.1 XP_016874584.1 13 1370
GenBank mRNA AB023163.1 - 17 1922
AB024494.1 - 17 2468
AB097013.1 - 17 1798
AB097035.1 - 17 1798
AK292866.1 - 17 1794
BC050324.1 - 17 1937 5266097

View Full Product Details