Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr. 1: 16985958 - 17011972 on Build GRCh38
UniGene ID
Hs.128866
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001141973.2 NP_001135445.1 8 882
NM_001141974.2 NP_001135446.1 8 882
NM_022089.3 NP_071372.1 8 897
XM_005245810.1 XP_005245867.1 8 894
XM_005245811.1 XP_005245868.1 8 882
XM_005245812.1 XP_005245869.1 8 897
XM_005245815.1 XP_005245872.1 8 897
XM_006710512.1 XP_006710575.1 8 879
XM_006710513.1 XP_006710576.1 8 882
XM_011541128.1 XP_011539430.1 8 897
XM_011541129.1 XP_011539431.1 8 897
XM_017000844.1 XP_016856333.1 8 897
XM_017000845.1 XP_016856334.1 8 879
XM_017000846.1 XP_016856335.1 8 882
XM_017000847.1 XP_016856336.1 8 894
XM_017000848.1 XP_016856337.1 8 897
XM_017000849.1 XP_016856338.1 8 882
XM_017000850.1 XP_016856339.1 8 897
GenBank mRNA AK290210.1 - 8 838
AL354615.1 - 8 697
AY461712.1 - 8 696
BC030267.1 - 8 825 5240813
BC034575.1 - 5 388

View Full Product Details