Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271684.1 | NP_001258613.1 | 5 | 872 | |
NM_001271685.1 | NP_001258614.1 | 5 | 707 | ||
NM_012243.2 | NP_036375.1 | 5 | 872 | ||
XM_005270691.4 | XP_005270748.1 | 6 | 1393 | ||
XM_011541135.2 | XP_011539437.1 | 4 | 726 | ||
XM_011541136.2 | XP_011539438.1 | 6 | 789 | ||
XM_011541137.2 | XP_011539439.1 | 5 | 849 | ||
XM_011541138.2 | XP_011539440.1 | 5 | 858 | ||
XM_017000869.1 | XP_016856358.1 | 4 | 1171 | ||
XM_017000870.1 | XP_016856359.1 | 5 | 1294 | ||
XM_017000871.1 | XP_016856360.1 | 4 | 1171 | ||
XM_017000872.1 | XP_016856361.1 | 4 | 1139 | ||
GenBank mRNA | AB021981.1 | - | 5 | 677 | |
AK290573.1 | - | 5 | 630 | ||
BC005136.2 | - | 5 | 655 | 3505772 | |
CR749816.1 | - | 5 | 707 |