Gene Transcripts
- Gene Symbol
- CDK20
- Entrez Gene ID
- 23552
- Gene Name
- cyclin dependent kinase 20
- Gene Aliases
- CCRK, CDCH, P42, PNQALRE
- Chromosome Location
- Chr. 9: 87966441 - 87974780 on Build GRCh38
- UniGene ID
- Hs.522274
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039803.2 |
NP_001034892.1
|
2, 3 |
518 |
|
NM_001170639.1 |
NP_001164110.1
|
2, 3 |
518 |
|
NM_001170640.1 |
NP_001164111.1
|
2, 3 |
518 |
|
NM_012119.4 |
NP_036251.2
|
2, 3 |
518 |
|
NM_178432.3 |
NP_848519.1
|
2, 3 |
557 |
|
XM_017014561.1 |
XP_016870050.1
|
2, 3 |
448 |
|
XM_017014562.1 |
XP_016870051.1
|
2, 3 |
448 |
|
XM_017014563.1 |
XP_016870052.1
|
2, 3 |
448 |
|
XM_017014564.1 |
XP_016870053.1
|
2, 3 |
448 |
|
GenBank mRNA |
AF035013.1 |
- |
2, 3 |
421 |
|
AF113130.1 |
- |
2, 3 |
429 |
|
AK075325.1 |
- |
2, 3 |
310 |
|
AK226135.1 |
- |
1, 2 |
608 |
|
AK291713.1 |
- |
2, 3 |
439 |
|
AK294193.1 |
- |
2, 3 |
271 |
|
AK296328.1 |
- |
2, 3 |
269 |
|
AK298993.1 |
- |
2, 3 |
261 |
|
AY904367.1 |
- |
2, 3 |
184 |
|
BC002655.2 |
- |
2, 3 |
243 |
3605918 |
BF966262.1 |
- |
2, 3 |
496 |
|
BI757318.1 |
- |
2, 3 |
369 |
|
BX647489.1 |
- |
2, 3 |
447 |
|
CB992644.1 |
- |
2, 3 |
527 |
|
KU178572.1 |
- |
2, 3 |
184 |
|
KU178573.1 |
- |
2, 3 |
184 |
|
KU178574.1 |
- |
2, 3 |
184 |
|
KU178575.1 |
- |
2, 3 |
184 |
|
KU178576.1 |
- |
2, 3 |
184 |
|
KU178577.1 |
- |
2, 3 |
184 |
|
View Full Product Details